Learn More
Selleck Chemical LLC Cabazitaxel S3022-10mM/1mL
SDP
Catalog No. 503108112
Encompass_Preferred
Cabazitaxel is a semi-synthetic derivative of a natural taxoid that kills cancer cells by inhibiting cell division and growth Cabazitaxel exerts its effects by inhibiting microtubule growth and assembly processes that are essential for cells to divide Cabazitaxel induces autophagy via the PI3K/Akt/mTOR pathway
Catalog No. 50-310-8112 Supplier Selleck Chemical LLC Supplier No. S302210MM/1ML
Cabazitaxel is a semi-synthetic derivative of a natural taxoid that kills cancer cells by inhibiting cell division and growth Cabazitaxel exerts its effects by inhibiting microtubule growth and assembly processes that are essential for cells to divide Cabazitaxel induces autophagy via the PI3K/Akt/mTOR pathway