Learn More
Lkt Laboratories Cabazitaxel, ≥98%, 10MG
SDP
Supplier:  Lkt Laboratories C00204110MG
Encompass_Preferred
Taxoid that disrupts the microtubular network in cells that exhibit Pgp mediated drug resistance. CAS: 183133-96-2. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-5028