Learn More
Medchemexpress LLC Cabazitaxel-d9 | 1383572-19-7 | C45H48D9NO14 | 1 MG
SDP
Catalog No. 5000376470
Encompass_Preferred
Cabazitaxel-d9 is deuterium labeled Cabazitaxel, a semi-synthetic derivative of the natural taxoid 10-deacetylbaccatin III with potential antineoplastic activity.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-76470 Supplier Medchemexpress LLC Supplier No. HY15459S11MG
Cabazitaxel-d9 is deuterium labeled Cabazitaxel, a semi-synthetic derivative of the natural taxoid 10-deacetylbaccatin III with potential antineoplastic activity.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs