Learn More
Cayman Chemical CabergolIn-d6 100ug
SDP
Catalog No. 502990928
Encompass_Preferred
An internal standard for the quantification of cabergoline by GC- or LC-MS
Catalog No. 50-299-0928 Supplier Cayman Chemical Supplier No. 31690100 UG
An internal standard for the quantification of cabergoline by GC- or LC-MS