Learn More
Medchemexpress LLC Cabergoline-d5 | 1426173-20-7 | C26H32D5N5O2 | 250 UG
SDP
Catalog No. 5000374485
Encompass_Preferred
Cabergoline-d5 is the deuterium-labeled version of Cabergoline, an ergot-derived dopamine D2-like receptor agonist with high affinity for D2, D3, and 5-HT2B receptors. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Can be utilized as a tracer in research applications
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
  • Deuteration can affect pharmacokinetic and metabolic profiles
  • Intended for research purposes only
Catalog No. 50-003-74485 Supplier Medchemexpress LLC Supplier No. 250UGHY15296S
Cabergoline-d5 is the deuterium-labeled version of Cabergoline, an ergot-derived dopamine D2-like receptor agonist with high affinity for D2, D3, and 5-HT2B receptors. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Can be utilized as a tracer in research applications
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
  • Deuteration can affect pharmacokinetic and metabolic profiles
  • Intended for research purposes only