Learn More
Chemglass Life Sciences Cabinet, Dry-Keeper™, Electric, Desiccator
SDP
Catalog No. 50401780
Encompass_Preferred
Cabinet, Dry-Keeper™, Electric, Desiccator. Durable desiccator cabinet has clear PVC panels and sleek black ABS accents, vertical profile, and movable shelves for adjustable dry storage configurations. Part number CG-1225-11 is equipped with a 110V, 6-watt regeneration unit for the silica gel. The gel is contained in the control section, where it absorbs moisture to maintain a low (30-40%) relative humidity. The absorption period lasts 4 1/2 hours, indicated by a green pilot light.
Catalog No. 50-401-780 Supplier Chemglass Life Sciences Supplier No. CG122511
Cabinet, Dry-Keeper™, Electric, Desiccator. Durable desiccator cabinet has clear PVC panels and sleek black ABS accents, vertical profile, and movable shelves for adjustable dry storage configurations. Part number CG-1225-11 is equipped with a 110V, 6-watt regeneration unit for the silica gel. The gel is contained in the control section, where it absorbs moisture to maintain a low (30-40%) relative humidity. The absorption period lasts 4 1/2 hours, indicated by a green pilot light.