Learn More
Ampco Cabinet-Tip Screwdriver
SDP
Catalog No. 19174282 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
17 in.
6 -5/8 in.
7 -5/8 in.
9 -1/2 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-174-282 7 -5/8 in.
19-174-283 6 -5/8 in.
19-174-285 9 -1/2 in.
19-174-286 17 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-174-282 Supplier Ampco Supplier No. S52
Add to Cart
Add to Cart
Cabinet-tip screwdriver with cellulose acetate handle and fluted grip. Non-sparking, non-magnetic, and corrosion-resistant with lifetime warranty.
  • Slotted tip
  • Fluted grip
  • Beryllium copper material
  • Lifetime warranty
  • Made in the USA
Specifications
Product Type Cabinet-Tip Screwdriver
Length (English) 7 -5/8 in.
Tip Style 1/4 in. Slotted