Learn More
World Precision Instrument Rechargeable Battery Pack for EVOM2
SDP
Catalog No. NC0180392
Encompass
The EVOM2 is equipped with a 6V 2700mAh rechargeable NiMH battery pack which can be repeatedly charged and discharged for approximately 500 cycles. The battery pack may be charged at any time in the discharge cycle, and can be charged continuously using the EVOM2 charger without damage.
Catalog No. NC0180392 Supplier World Precision Instrument Supplier No. 91736
The EVOM2 is equipped with a 6V 2700mAh rechargeable NiMH battery pack which can be repeatedly charged and discharged for approximately 500 cycles. The battery pack may be charged at any time in the discharge cycle, and can be charged continuously using the EVOM2 charger without damage.