Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Cable Assembly for Remote Control, 18-pin Round Connector; 25 ft (7.6 m) Cable Length
Catalog No. NC0701678
Encompass
Cables include noted terminal connector and cable with stripped wire ends for connection to PLC or other control interface.
Catalog No. NC0701678 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7730032
Cables include noted terminal connector and cable with stripped wire ends for connection to PLC or other control interface.