Learn More
Cole-Parmer Cole-Parmer Electrode cable, detachable, 10-ft, BNC
Catalog No. 50633599 Shop All Cole-Parmer Products
Encompass_Preferred
Detachable cable features BNC connection for compatibility with most pH meters and controllers.
Catalog No. 50-633-599 Supplier Cole-Parmer Supplier No. 0566256
Detachable cable features BNC connection for compatibility with most pH meters and controllers.