Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Cable 2 Conduct 22AWG w/Shield

Catalog No. 5000207869 Shop All Hach Company Products
Encompass

Cable-2 Conductors 22 AWG with Shield

Catalog No. 50-002-07869 Supplier Hach Company Supplier No. 9707200
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cable-2 Conductors 22 AWG with Shield

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.