Learn More
Cole-Parmer Cole-Parmer Electrode cable, detachable, 3-ft, BNC
Supplier:  Cole-Parmer 0566255
Encompass_Preferred
Detachable cable features BNC connection for compatibility with most pH meters and controllers.
Catalog No. 50-633-598