Learn More
EZKEM Cable Amperometric Detector for the FS3700
SDP
Catalog No. NC1813188
Encompass_Preferred
EZkem’s Amperometric Detector cable for the FS3700 instrument is available as a replacement cable for a worn cable or necessary if you are adding Cyanide (and our Amperometric Flow Cell) to a instrument already running a photometric channel (or if you are converting a photometric channel to an ampermetric channel).
Catalog No. NC1813188 Supplier EZKEM Supplier No. 330061
EZkem’s Amperometric Detector cable for the FS3700 instrument is available as a replacement cable for a worn cable or necessary if you are adding Cyanide (and our Amperometric Flow Cell) to a instrument already running a photometric channel (or if you are converting a photometric channel to an ampermetric channel).