Learn More
Waters Corp Cable Assembly, Sample Manager to Column Heater, UPLC, ACQUITY Compatible
Catalog No. 502392603 Shop All Waters Corporation Products
Encompass_Preferred
This cable assembly is a replacement component designed to connect a Sample Manager to a Column Heater or Column Heater Cooler. It is specifically compatible with ACQUITY Sample Managers and 30cm Column Heater Modules, operating within UPLC systems.
  • Connects sample manager to column heater/cooler
  • Compatible with ACQUITY sample managers
  • Compatible with 30cm column heater modules
  • Designed for UPLC systems
  • Provided as a spare parts kit
  • Mechanical type
  • Replaces as required
Catalog No. 50-239-2603 Supplier Waters Corp Supplier No. 700011057
This cable assembly is a replacement component designed to connect a Sample Manager to a Column Heater or Column Heater Cooler. It is specifically compatible with ACQUITY Sample Managers and 30cm Column Heater Modules, operating within UPLC systems.
  • Connects sample manager to column heater/cooler
  • Compatible with ACQUITY sample managers
  • Compatible with 30cm column heater modules
  • Designed for UPLC systems
  • Provided as a spare parts kit
  • Mechanical type
  • Replaces as required