Learn More
Hach Company Thermistor Cable Assembly for Reagent Sensor (Replacement for Hach Instruments)
Catalog No. NC3304037 Shop All Hach Company Products
Encompass
Replacement thermistor cable assembly designed to connect the thermistor element in reagent sensor assemblies and provide temperature sensing or compensation for reagent-based measurements. Use as a spare or replacement cable to restore thermistor connectivity and support accurate temperature-dependent readings.
  • Provides thermistor connection for reagent sensor temperature measurement.
  • Used as a spare or replacement cable for compatible reagent sensor assemblies.
  • Facilitates accurate temperature compensation for reagent-based measurements.
  • Helps maintain instrument performance by replacing damaged or worn cables.
Catalog No. NC3304037 Supplier Hach Company Supplier No. 6779100
Replacement thermistor cable assembly designed to connect the thermistor element in reagent sensor assemblies and provide temperature sensing or compensation for reagent-based measurements. Use as a spare or replacement cable to restore thermistor connectivity and support accurate temperature-dependent readings.
  • Provides thermistor connection for reagent sensor temperature measurement.
  • Used as a spare or replacement cable for compatible reagent sensor assemblies.
  • Facilitates accurate temperature compensation for reagent-based measurements.
  • Helps maintain instrument performance by replacing damaged or worn cables.