Learn More
Hach Company Cable Assembly, Pinch Valve, 3-Way - Replacement for Hach 5500sc and 961Xsc Process Analyzers
Catalog No. NC2213425 Shop All Hach Company Products
Encompass
This cable assembly provides the electrical and mechanical connection to operate a 3-way pinch valve used for sample handling in process analyzers, commonly used in silica and phosphate analysis. It serves as a replacement cable for single-channel analyzer configurations and is intended for maintenance and repair of valve assemblies.
  • Connects and transmits control signals to a 3-way pinch valve used in sample handling.
  • Designed for use in single-channel process analyzers for silica and phosphate measurements.
  • Provides a direct replacement for factory-installed valve cable assemblies.
  • Simplifies maintenance and reduces analyzer downtime when replacing worn or damaged cables.
  • No dimensional specifications are listed on the manufacturer product page; confirm fit and connections before installation.
This item is not returnable. View return policy
Catalog No. NC2213425 Supplier Hach Company Supplier No. 6786500
This cable assembly provides the electrical and mechanical connection to operate a 3-way pinch valve used for sample handling in process analyzers, commonly used in silica and phosphate analysis. It serves as a replacement cable for single-channel analyzer configurations and is intended for maintenance and repair of valve assemblies.
  • Connects and transmits control signals to a 3-way pinch valve used in sample handling.
  • Designed for use in single-channel process analyzers for silica and phosphate measurements.
  • Provides a direct replacement for factory-installed valve cable assemblies.
  • Simplifies maintenance and reduces analyzer downtime when replacing worn or damaged cables.
  • No dimensional specifications are listed on the manufacturer product page; confirm fit and connections before installation.