Learn More
Hach Company Cable Assy Pwr Adpt 2 to 3 Pin
Supplier:  Hach Company 8739400
Encompass
Cable Assembly-Power Adaptor 2 to 3 Pin2 to 3 Pin Power Adaptor allows use of Prod No 1414 batteries with new 3 pin SD900 controller
Catalog No. 50-002-07102