Learn More
World Precision Instrument BNC-to-Mini Phone Plug Cable for DAM Series Extracellular Amplifiers
SDP
Catalog No. 50822142
Encompass
Our BNC-to-Mini Phone Plug Coaxial Cable is designed for use with the DAM series Amplifiers, like the DAM50 and the DAM80 Extracellular Amplifiers. It is 1.8m (6 feet) long to accommodate most electrophysiology applications in your laboratory. These cables have a metal, male BNC connector on one end of the cable and a 3.5mm mini phone plug on the other end. BNC (Bayonet Neill-Concelmen) connectors have a bayonet-style locking mechanism that provides a secure connection. These coaxial cables have an impedance of 50 ohms (RG58U style). The cables have excellent shielding from electromagnetic interference and radio frequency interference, which is perfect for electrophysiology laboratories. 
Catalog No. 50-822-142 Supplier World Precision Instrument Supplier No. CBL102
Our BNC-to-Mini Phone Plug Coaxial Cable is designed for use with the DAM series Amplifiers, like the DAM50 and the DAM80 Extracellular Amplifiers. It is 1.8m (6 feet) long to accommodate most electrophysiology applications in your laboratory. These cables have a metal, male BNC connector on one end of the cable and a 3.5mm mini phone plug on the other end. BNC (Bayonet Neill-Concelmen) connectors have a bayonet-style locking mechanism that provides a secure connection. These coaxial cables have an impedance of 50 ohms (RG58U style). The cables have excellent shielding from electromagnetic interference and radio frequency interference, which is perfect for electrophysiology laboratories. 