Learn More
METTLER TOLEDO  Analog Conductivity Patch Cable, 25 Foot, M300 Standard
Catalog No. NC1862666 Shop All METTLER TOLEDO Products
Encompass
*Mettler Toledo's Patch Cables are used for connections between sensors and M300 instruments.*H19:H20Length: 25 foot*Uses: Conductivity / Resistivity
Catalog No. NC1862666 Supplier METTLER TOLEDO Supplier No. 58080254
*Mettler Toledo's Patch Cables are used for connections between sensors and M300 instruments.*H19:H20Length: 25 foot*Uses: Conductivity / Resistivity