Learn More
MSC Crescent Cable Cutter Pliers, Alloy Steel, 8 in., 1 ea.
Catalog No. 5000507968
Encompass
These 8 in. cable cutters are designed for professionals in construction, electrical work, and telecommunications. Manufactured from alloy steel for enhanced strength and durability, these pliers feature precision sheer-cut blades that provide clean, sharp cuts with minimal cable distortion. They are optimized for cutting copper, aluminum, and communication cables up to 4-0 AWG.
  • Sheer-cut blades for clean, sharp results
  • Cutting capacity up to 0.43 in. (4-0 AWG)
  • Ergonomic dipped grips for comfort and easy cleaning
  • Meets or exceeds ASME and ANSI specifications
  • Full lifetime warranty
Catalog No. 50-005-07968 Supplier MSC Supplier No. 67895011
These 8 in. cable cutters are designed for professionals in construction, electrical work, and telecommunications. Manufactured from alloy steel for enhanced strength and durability, these pliers feature precision sheer-cut blades that provide clean, sharp cuts with minimal cable distortion. They are optimized for cutting copper, aluminum, and communication cables up to 4-0 AWG.
  • Sheer-cut blades for clean, sharp results
  • Cutting capacity up to 0.43 in. (4-0 AWG)
  • Ergonomic dipped grips for comfort and easy cleaning
  • Meets or exceeds ASME and ANSI specifications
  • Full lifetime warranty