Learn More
Cole-Parmer Cole-Parmer pH Electrode Extension Cable, 25 ft
Supplier:  Cole-Parmer 0599457
Encompass_Preferred
Heavy-duty shielded extension cables for pH and ORP electrodes are insulated for strength and noise reduction.
Catalog No. 50-633-605