Learn More
Cole-Parmer Cole-Parmer pH Electrode Extension Cable, 50 ft
Catalog No. 50633606 Shop All Cole-Parmer Products
Encompass_Preferred
Heavy-duty shielded extension cables for pH and ORP electrodes are insulated for strength and noise reduction.
Catalog No. 50-633-606 Supplier Cole-Parmer Supplier No. 0599458
Heavy-duty shielded extension cables for pH and ORP electrodes are insulated for strength and noise reduction.