Learn More
Metrohm USA CABLE GEN ELEC 6.2104.120
SDP
Catalog No. NC0418618 Shop All Metrohm USA Products
Encompass
With plug H. For connecting generator electrode - KF Coulometer.
Catalog No. NC0418618 Supplier Metrohm USA Supplier No. 62104120
With plug H. For connecting generator electrode - KF Coulometer.