Learn More
Grainger Ohaus RS232 Cable, IBM 9P Male-to-Female, 72 in., 1 ea.
Catalog No. 502768960
Encompass_Preferred
This RS232 cable facilitates communication between compatible weighing instruments and IBM 9-pin PC systems. Designed for reliable data transfer, this accessory ensures secure connectivity for various measurement and processing applications.
  • Features an IBM 9-pin male-to-female connector configuration
  • Measures 72 in. in overall length
  • Constructed with durable gray plastic coated wire
  • Compatible with Adventure Precision, Defenders, Rangers 3000 and 4000, Valors, and Explorers models
Catalog No. 50-276-8960 Supplier Grainger Supplier No. 13P637
This RS232 cable facilitates communication between compatible weighing instruments and IBM 9-pin PC systems. Designed for reliable data transfer, this accessory ensures secure connectivity for various measurement and processing applications.
  • Features an IBM 9-pin male-to-female connector configuration
  • Measures 72 in. in overall length
  • Constructed with durable gray plastic coated wire
  • Compatible with Adventure Precision, Defenders, Rangers 3000 and 4000, Valors, and Explorers models