Learn More
Grainger Ohaus Printer Cable, Plastic Coated Wire, 1 ea
Catalog No. 502768874
Encompass_Preferred
The ST103-EX MB PA TxxP cable provides a reliable interface for data transmission between OHAUS weighing balances and compatible thermal printers. This printer cable is designed to support seamless communication, facilitating efficient printing processes for various laboratory and industrial weighing applications.
  • Features a plastic coated wire construction for durability
  • Compatible with a wide range of OHAUS equipment including Adventurer, Explorer, Pioneer, and various moisture analyzer series
  • Designed for straightforward RS232 connectivity
  • Facilitates printing functionality between peripheral devices
Catalog No. 50-276-8874 Supplier Grainger Supplier No. 13P635
The ST103-EX MB PA TxxP cable provides a reliable interface for data transmission between OHAUS weighing balances and compatible thermal printers. This printer cable is designed to support seamless communication, facilitating efficient printing processes for various laboratory and industrial weighing applications.
  • Features a plastic coated wire construction for durability
  • Compatible with a wide range of OHAUS equipment including Adventurer, Explorer, Pioneer, and various moisture analyzer series
  • Designed for straightforward RS232 connectivity
  • Facilitates printing functionality between peripheral devices