Learn More
Molecular Devices Molecular Devices Waste Reservoir, 10 L, Plastic, Replacement
Catalog No. 5000014564
Encompass_Preferred
The Molecular Devices 10 L AquaMax waste reservoir is a replacement waste collection bottle for AquaMax microplate washers (models 2000 and 4000). It provides 10 liters of capacity and connects via the supplied cap and tubing for efficient waste handling during plate-washing operations.
  • 10 L capacity for extended plate-washing runs.
  • Supplied with cap, fluid tubing, and waste tubing for straightforward installation.
  • Plastic construction; inspect and replace when signs of aging appear.
  • Designed for laboratory waste collection during microplate washing workflows.
  • Supports standard bottle connections used with microplate washer accessories.
Catalog No. 50-000-14564 Supplier Molecular Devices Supplier No. 90000296
The Molecular Devices 10 L AquaMax waste reservoir is a replacement waste collection bottle for AquaMax microplate washers (models 2000 and 4000). It provides 10 liters of capacity and connects via the supplied cap and tubing for efficient waste handling during plate-washing operations.
  • 10 L capacity for extended plate-washing runs.
  • Supplied with cap, fluid tubing, and waste tubing for straightforward installation.
  • Plastic construction; inspect and replace when signs of aging appear.
  • Designed for laboratory waste collection during microplate washing workflows.
  • Supports standard bottle connections used with microplate washer accessories.