Learn More
World Precision Instrument Low-noise cable for Microelectrode Holder, A : pin, B : pin
SDP
Catalog No. NC0463483
Encompass
  • 2mm pin-socket to 2mm pin-socket or 2 mm tip-plug stackable
  • Cable: 2 ft (0.6 m)
Catalog No. NC0463483 Supplier World Precision Instrument Supplier No. 5371
  • 2mm pin-socket to 2mm pin-socket or 2 mm tip-plug stackable
  • Cable: 2 ft (0.6 m)