Learn More
World Precision Instrument 2 Optional Shielded Modular Cables for DAM50 Bioamplifier
SDP
Catalog No. 50822066
Encompass
Shielded cable for DAM50
This shielded cable is often used with the WPI DAM50 Bioamplifier. It has an RJ11 connector  for the DAM50 input port. The other end is unterminated so you can build the piece you need for you own application. 
Catalog No. 50-822-066 Supplier World Precision Instrument Supplier No. 3517
Shielded cable for DAM50
This shielded cable is often used with the WPI DAM50 Bioamplifier. It has an RJ11 connector  for the DAM50 input port. The other end is unterminated so you can build the piece you need for you own application. 