Learn More
Bellco Glass, Inc. Cable pH BNC TO HP,6' Broadley-James
SDP
Catalog No. 501946303
Encompass_Preferred
The Fermprobe pH electrode with S8 metric cap features a standard S8 detachablecable connector on a metric threaded polymer cap. The design requires the rotation of the electrode during installation. This style of pH Fermprobe is the firstchoice for small autoclavable bench-top vessels. All Fermprobe pH electrodes are designed to be used in bio-process applications where clean in place sterilize in place CIP, SIP procedures are used. Built to withstand repeated steam sterilization cycles the Fermprobe quickly stabilizes after steam exposure to be on line in half the time of other electrodes. This rugged low-impedance pH electrode continues to provide fast and precise measurement.
Catalog No. 50-194-6303 Supplier Bellco Glass, Inc. Supplier No. 780322550
The Fermprobe pH electrode with S8 metric cap features a standard S8 detachablecable connector on a metric threaded polymer cap. The design requires the rotation of the electrode during installation. This style of pH Fermprobe is the firstchoice for small autoclavable bench-top vessels. All Fermprobe pH electrodes are designed to be used in bio-process applications where clean in place sterilize in place CIP, SIP procedures are used. Built to withstand repeated steam sterilization cycles the Fermprobe quickly stabilizes after steam exposure to be on line in half the time of other electrodes. This rugged low-impedance pH electrode continues to provide fast and precise measurement.