Learn More
Werner Co. Cable Positioning Lanyard
Catalog No. 17103814
Change view
Click to view available options
Material:
Steel
Catalog No. Material
17-103-814 Steel
1 options
Catalog No. 17-103-814 Supplier Werner Co. Supplier No. C161106
  • $98.00
    $90.65 / Each of 1
    Save $7.35
    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Cable positioning lanyard with snap hook.
Recommended for work positioning and restraint applications only.
  • Vinyl coated 1/4 in. cable is heat resistant and highly abrasion resistant.
  • Snap hook is designed to prevent false engagement when connecting to the harness.
Specifications
Material Steel
Connector Types Snap Hook
Includes Snap Hook
Color Silver
Certifications/Compliance ANSI Z359.3-2019
Leg Configuration Single
Length (English) 6 ft.
Length (Metric) 1.83 m
Product Type Positioning Lanyard