Learn More
World Precision Instrument Cable, shielded transducer stock
SDP
Supplier:  World Precision Instrument 5385
Encompass
Popular electrical component
This shielded transducer stock cable has no connectors on either end. it is 25 ft (7.6 m) long. It has 8 conductor, 30 gauge stranded wires with shield cable, and it is Teflon insulated.
Catalog No. 50-822-113