Learn More
Leica Biosystems Shutter-control cable for Leica EL6000 (I²C)
Catalog No. NC3366007
Encompass_Preferred
Leica Microsystems part 11500336 is a shutter-control cable for the EL6000 external light source. It provides I²C-based shutter signaling to operate the EL6000's integrated fast shutter, enabling fast on/off control of excitation light for fluorescence imaging. Intended as a spare or replacement accessory for EL6000-equipped microscope systems.
  • Provides shutter control via I²C interface.
  • Designed for use with EL6000 external fluorescence light sources.
  • Enables fast shutter operation for timed excitation sequences.
  • Intended as a spare or replacement accessory.
  • Requires compatible connector and control system.
Catalog No. NC3366007 Supplier Leica Biosystems Supplier No. 11500336
Leica Microsystems part 11500336 is a shutter-control cable for the EL6000 external light source. It provides I²C-based shutter signaling to operate the EL6000's integrated fast shutter, enabling fast on/off control of excitation light for fluorescence imaging. Intended as a spare or replacement accessory for EL6000-equipped microscope systems.
  • Provides shutter control via I²C interface.
  • Designed for use with EL6000 external fluorescence light sources.
  • Enables fast shutter operation for timed excitation sequences.
  • Intended as a spare or replacement accessory.
  • Requires compatible connector and control system.