Learn More
Grainger Cable Tie 3/8 W Cable Tie 2 Max OD PK50
Supplier:  Grainger 16P344
Encompass_Preferred
The Grainger Approved Cable Tie is a 4-inch long, 0.12-inch wide nylon cable tie that can bundle and secure wires and cables. It has a minimum loop tensile strength of 18 lbs and a maximum bundle diameter of 1.18 inches. The cable tie has a one-piece construction with a smooth tip and secure locking feature. It comes in black color and is rated for indoor and outdoor use. The operating temperature range is -40°F to 185°F. It meets military, aerospace, and commercial specifications. This cable tie provides a cost-effective solution for bundling and securing wires and cables.
Catalog No. 50-276-5947