Learn More
Grainger Cable Tie Gun HD 100 lb SS
Catalog No. 502749406
Encompass_Preferred
The Dolphin Components Corp Cable Tie Tension Tool For Nylon Cable Ties is a handheld tensioning tool designed to tighten nylon cable ties to a specific tension. It features an ergonomic soft grip handle and a tension adjustment knob that allows the user to dial in the desired tension from 15 to 80 pounds. The tool has a one-handed squeeze operation that uniformly tensions and cuts off the cable tie tail in one motion. It accepts cable tie widths from .08" to .23" and lengths up to 18". This tool helps ensure proper cable tie application and provides cable bundles with consistent, uniform tension.
Catalog No. 50-274-9406 Supplier Grainger Supplier No. 14L582
The Dolphin Components Corp Cable Tie Tension Tool For Nylon Cable Ties is a handheld tensioning tool designed to tighten nylon cable ties to a specific tension. It features an ergonomic soft grip handle and a tension adjustment knob that allows the user to dial in the desired tension from 15 to 80 pounds. The tool has a one-handed squeeze operation that uniformly tensions and cuts off the cable tie tail in one motion. It accepts cable tie widths from .08" to .23" and lengths up to 18". This tool helps ensure proper cable tie application and provides cable bundles with consistent, uniform tension.