Learn More
Chemglass Life Sciences Cable Tie Gun, Plastic, Adjustable Tension
SDP
Catalog No. 501532117
Encompass
MATERIAL: Plastic.COMPATIBILITY: Accommodates cable ties up to19" in width.Not compatible with stainless steel ties.FEATURES: Produces 18-50lbs of tensile strength Gun tightens and cuts.
Catalog No. 50-153-2117 Supplier Chemglass Life Sciences Supplier No. CLS1399G100
MATERIAL: Plastic.COMPATIBILITY: Accommodates cable ties up to19" in width.Not compatible with stainless steel ties.FEATURES: Produces 18-50lbs of tensile strength Gun tightens and cuts.