Learn More
Grainger Plastic Cable Tie Tension Tool for Nylon, Ergonomic Handle, 8 to 50 lb.
Catalog No. NC1705209
Encompass_Preferred
Trigger-handle Standard-Duty Cable Tie Tool can be used to install subminiature, miniature, intermediate, and standard cross-section cable ties. Adjustable tension. Requires minimal handle force to cut off the cable tie. Gray impact-resistant resin housing and a comfortable, ergonomically designed black cushion handle that minimizes hand impact.
Catalog No. NC1705209 Supplier Grainger Supplier No. 52ZJ96
Trigger-handle Standard-Duty Cable Tie Tool can be used to install subminiature, miniature, intermediate, and standard cross-section cable ties. Adjustable tension. Requires minimal handle force to cut off the cable tie. Gray impact-resistant resin housing and a comfortable, ergonomically designed black cushion handle that minimizes hand impact.