Learn More
Andwin Scientific CABLE TIES, 100/PK
SDP
Supplier:  Andwin Scientific 89030224
Encompass_Preferred
The cable ties can be utilized to hang equipments and bundle materials. With tapered tips, these ties can be self-locked, quickly insertened, and tightened. This product can withstand temperature range of -40 to 85�C (-40 to 185�F) and resist chemical and corrosion. The ties are also rustproof and nonconducting. It comes in a pack of 100.
Catalog No. NC0494774