Learn More
Andwin Scientific CABLE TIES 11IN 100PK
SDP
Catalog No. NC9577181
Encompass_Preferred
The 11" nylon plastic cable ties can fasten anything up to 50 pounds or bundles with a diameter of 1/16" to 3". These ties are long-lasting and are UL approved. The nylon is classified as type 6.6. This product is sold in a pack of 100.
Catalog No. NC9577181 Supplier Andwin Scientific Supplier No. 95481KY
The 11" nylon plastic cable ties can fasten anything up to 50 pounds or bundles with a diameter of 1/16" to 3". These ties are long-lasting and are UL approved. The nylon is classified as type 6.6. This product is sold in a pack of 100.