Learn More
Lovibond™ Cable, USB-C
Catalog No. 09054118 Shop All Tintometer Group Products
Change view
Click to view available options
For Use With (Equipment):
Turb 350/355 IR and 355 T Portable Turbidimeter
Catalog No. For Use With (Equipment)
09-054-118 Turb 350/355 IR and 355 T Portable Turbidimeter
1 options
This item is not returnable. View return policy
Catalog No. 09-054-118 Supplier Lovibond™ Supplier No. 19820081
Add to Cart
Add to Cart
This item is not returnable. View return policy
A standard industry USB-C Cable that is able to deliver power to the Lovibond™ TB Series of portable turbidimeters when combined with a power supply.
Specifications
Type USB-C Cable
For Use With (Equipment) Turb 350/355 IR and 355 T Portable Turbidimeter
For Use With (Application) Alternate power supply option for Lovibond TB350 turbidimeters.
Length (English) 3.3 ft.
Length (Metric) 1 m