Learn More
Warner Instruments™ Cable With Bead Thermistor
Catalog No. BTX640107 Shop All BTX Harvard Products
Encompass_Preferred
Change view
Click to view available options
Temperature (Metric):
5°C to 40°C
Catalog No. Temperature (Metric)
BTX640107 5°C to 40°C
1 options
Catalog No. BTX640107 Supplier Warner Instruments™ Supplier No. 640107
Cable with a 0.85 x 4.6 mm bead thermistor and BNC end is used with most in-line solution heaters and accepts the TS-70B replacement.
  • Interface cables and thermisitors for connecting Warner temperature controllers to Warner products
Specifications
Connectors BNC
Voltage 0/1 V
Model TA-29
Temperature (Metric) 5°C to 40°C