Learn More
Perkin Elmer US LLC Non-Coded Lumina Lamp Plug Adapter
Catalog No. 509044272
Encompass_Preferred
Allows Lumina™ HCLs (N305XXXX series, 4-pin) to be used with PerkinElmer atomic absorption instruments with non-coded lamp connectors (9-pin).
Catalog No. 50-904-4272 Supplier Perkin Elmer US LLC Supplier No. N3050198
Allows Lumina™ HCLs (N305XXXX series, 4-pin) to be used with PerkinElmer atomic absorption instruments with non-coded lamp connectors (9-pin).