Learn More
Aviva Systems Biology Corp CABLES1 Antibody - C-terminal region (ARP64759_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64759_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CABLES1. It was validated on Western Blot. . Alternate names: CABLES, FLJ35924, HsT2563, IK3-1, CABL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-360