Learn More
TARGETMOL CHEMICALS INC CABOTEGRAVIR 50MG
SDP
Catalog No. 502579564
Encompass_Preferred
Also available in 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cabotegravir (S/GSK1265744) (GSK744 GSK1265744) is a long-acting HIV integrase inhibitor and inhibits the HIV-1 integrase catalyzed strand transfer reaction (IC50 3.0 nM). purity: 99%
Catalog No. 50-257-9564 Supplier TARGETMOL CHEMICALS INC Supplier No. T609850MG
Also available in 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cabotegravir (S/GSK1265744) (GSK744 GSK1265744) is a long-acting HIV integrase inhibitor and inhibits the HIV-1 integrase catalyzed strand transfer reaction (IC50 3.0 nM). purity: 99%