Learn More
Medchemexpress LLC HY-15592 50mg , Cabotegravir CAS:1051375-10-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1559250MG
Encompass_Preferred
HY-15592 50mg Cabotegravir CAS: 1051375-10-0 Cabotegravir (GSK-1265744) is a orally active and long-acting HIV integrase inhibitor and organic anion transporter 1/3 (OAT1/OAT3) inhibitor with IC50 values of 2.5 nM, 0.41 μM and 0.81 μM for HIVADA, OAT3 and OAT1, respectively. Cabotegravir is primarily metabolized by uridine diphosphate glucuronosyltransferase (UGT) 1A1, with low potential to interact with other antiretroviral drugs (ARVs). Cabotegravir can be used to research AIDS. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0170