Learn More
Cayman Chemical Cabotegravir, Purity: ≥98%, CAS: 1051375-10-0, 5 mg
SDP
Catalog No. 502940372
Encompass_Preferred
An HIV-1 integrase inhibitor (IC50 = 3 nM in a strand transfer assay); inhibits HIV replication in MT-4 cells and PBMCs (IC50s = 1.3 and 0.2 nM, respectively); protective against SHIV infection in macaques at 50 mg/kg
Catalog No. 50-294-0372 Supplier Cayman Chemical Supplier No. 272155 MG
An HIV-1 integrase inhibitor (IC50 = 3 nM in a strand transfer assay); inhibits HIV replication in MT-4 cells and PBMCs (IC50s = 1.3 and 0.2 nM, respectively); protective against SHIV infection in macaques at 50 mg/kg