Learn More
Selleck Chemical LLC Cabotegravir5mg
SDP
Supplier:  Selleck Chemical LLC S77665MG
Encompass_Preferred
Cabotegravir (GSK744, GSK1265744, S/GSK1265744) is a long-acting HIV integrase inhibitor against a broad range of HIV subtypes, and inhibits the HIV-1 integrase catalyzed strand transfer reaction with IC50 of 3.0 nM. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3304