Learn More
Enzo Life Sciences Cabozantinib (10mg). CAS: 849217-68-1
SDP
Catalog No. 502013496
Encompass_Preferred
Cabozantinib is a very potent inhibitor, with an IC50 of less than 10 nM for VEGFR2, MET, Kit and FLT3-ITD. Alternative name: XL184, BMS-907351, N-(4-((6,7-Dimethoxyquinolin-4-yl)oxy)phenyl)-N-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO (100mg/ml) or ethanol (1mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-3496 Supplier Enzo Life Sciences Supplier No. ENZCHM1820010
Cabozantinib is a very potent inhibitor, with an IC50 of less than 10 nM for VEGFR2, MET, Kit and FLT3-ITD. Alternative name: XL184, BMS-907351, N-(4-((6,7-Dimethoxyquinolin-4-yl)oxy)phenyl)-N-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO (100mg/ml) or ethanol (1mg/ml). Long Term Storage: -20°C.