Learn More
Medchemexpress LLC HY-13016 10mg , Cabozantinib CAS:849217-68-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1301610MG
Encompass_Preferred
HY-13016 10mg Cabozantinib CAS: 849217-68-1 Cabozantinib is a potent and orally active inhibitor of VEGFR2 and MET, with IC50 values of 0.035, and 1.3 nM, respectively. Cabozantinib displays strong inhibition of KIT, RET, AXL, TIE2, and FLT3 (IC50=4.6, 5.2, 7, 14.3, and 11.3 nM, respectively). Cabozantinib shows antiangiogenic activity. Cabozantinib disrupts tumor vasculature and promotes tumor and endothelial cell apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9076