Learn More
TARGETMOL CHEMICALS INC CABOZANTINIB 200MG
SDP
Catalog No. 502580735
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cabozantinib (BMS-907351) (XL184) is a potent pan-tyrosine kinases inhibitor that inhibits VEGFR2 c-Met Kit Axl and Flt3 (IC50s 0.035 1.3 4.6 7 and 11.3 nM). purity: 99%
Catalog No. 50-258-0735 Supplier TARGETMOL CHEMICALS INC Supplier No. T2586200MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cabozantinib (BMS-907351) (XL184) is a potent pan-tyrosine kinases inhibitor that inhibits VEGFR2 c-Met Kit Axl and Flt3 (IC50s 0.035 1.3 4.6 7 and 11.3 nM). purity: 99%