Learn More
Medchemexpress LLC Cabozantinib (Standard) | 849217-68-1 | 99.9% | 100 MG
SDP
Catalog No. 501637882
Encompass_Preferred
Cabozantinib is a potent and orally active inhibitor of VEGFR2 and MET. It also strongly inhibits KIT, RET, AXL, TIE2, and FLT3. Cabozantinib exhibits antiangiogenic activity, disrupting tumor vasculature and promoting apoptosis in tumor and endothelial cells.
  • Potent and orally active inhibitor
  • Inhibits VEGFR2 and MET
  • Strongly inhibits KIT, RET, AXL, TIE2, and FLT3
  • Exhibits antiangiogenic activity
  • Disrupts tumor vasculature
  • Promotes apoptosis in tumor and endothelial cells
Catalog No. 50-163-7882 Supplier Medchemexpress LLC Supplier No. HY13016100MG
Cabozantinib is a potent and orally active inhibitor of VEGFR2 and MET. It also strongly inhibits KIT, RET, AXL, TIE2, and FLT3. Cabozantinib exhibits antiangiogenic activity, disrupting tumor vasculature and promoting apoptosis in tumor and endothelial cells.
  • Potent and orally active inhibitor
  • Inhibits VEGFR2 and MET
  • Strongly inhibits KIT, RET, AXL, TIE2, and FLT3
  • Exhibits antiangiogenic activity
  • Disrupts tumor vasculature
  • Promotes apoptosis in tumor and endothelial cells